siRNA Details
| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145772.1 | NP_665715.1 | 4 | 495 | ||
| GenBank mRNA | AF181992.1 | - | 4 | 495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dync1li1| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145772.1 | NP_665715.1 | 4 | 495 | ||
| GenBank mRNA | AF181992.1 | - | 4 | 495 | ||