siRNA Details
| Gene Symbol: Ctsz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183330.1 | NP_899159.1 | 3 | 442 | ||
| GenBank mRNA | AB023781.2 | - | 3 | 442 | ||
| BC091110.1 | - | 3 | 376 | 7379777 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsz| Gene Symbol: Ctsz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183330.1 | NP_899159.1 | 3 | 442 | ||
| GenBank mRNA | AB023781.2 | - | 3 | 442 | ||
| BC091110.1 | - | 3 | 376 | 7379777 | ||