siRNA Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142366.2 | NP_001135838.1 | 2 | 233 | ||
| NM_001270558.2 | NP_001257487.1 | 2 | 225 | |||
| NM_001270559.2 | NP_001257488.1 | 2 | 233 | |||
| NM_012825.4 | NP_036957.1 | 2 | 225 | |||
| GenBank mRNA | EV243390.1 | - | 2 | 408 | ||
| EV243422.1 | - | 1 | 271 | |||
| EV243423.1 | - | 2 | 356 | |||
| L27588.1 | - | 1 | 93 | |||
| U14007.1 | - | 2 | 159 | |||

Aqp4
Genomic Map