siRNA Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164073.1 | NP_001157545.1 | 6 | 561 | ||
NM_145788.2 | NP_665731.2 | 6 | 572 | |||
XM_006234273.2 | XP_006234335.1 | 6 | 634 | |||
XM_006234274.2 | XP_006234336.1 | 6 | 620 | |||
GenBank mRNA | AJ422211.1 | - | 5 | 432 | ||
BC088854.1 | - | 7 | 640 | |||
BC129086.1 | - | 6 | 561 | 7365827 |
