siRNA Details
| Gene Symbol: Ghrhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012850.1 | NP_036982.1 | 12, 13 | 1183 | ||
| GenBank mRNA | AF122055.1 | - | 12, 13 | 1156 | ||
| AF409106.1 | - | 3, 4 | 355 | |||
| L01407.1 | - | 12, 13 | 1183 | |||
More Information
Set Membership: |
|

Ghrhr
Genomic Map