siRNA Details
| Gene Symbol: Asic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034014.1 | NP_001029186.1 | 2, 3 | 859 | ||
| NM_012892.2 | NP_037024.2 | 2, 3 | 874 | |||
| GenBank mRNA | U53211.1 | - | 2, 3 | 859 | ||
| Y14635.1 | - | 2, 3 | 874 | |||

Asic2
Genomic Map