siRNA Details
| Gene Symbol: Faslg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012908.1 | NP_037040.1 | 4 | 559 | ||
| XM_006250148.2 | XP_006250210.1 | 7 | 1198 | |||
| XM_008769607.1 | XP_008767829.1 | 8 | 1433 | |||
| GenBank mRNA | U03470.1 | - | 4 | 559 | ||

Faslg
Genomic Map