siRNA Details
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012914.1 | NP_037046.1 | 8 | 1074 | ||
XM_006246597.2 | XP_006246659.1 | 9 | 1688 | |||
GenBank mRNA | M30581.1 | - | 8 | 1074 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012914.1 | NP_037046.1 | 8 | 1074 | ||
XM_006246597.2 | XP_006246659.1 | 9 | 1688 | |||
GenBank mRNA | M30581.1 | - | 8 | 1074 |