siRNA Details
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033715.1 | NP_001028887.1 | 9 | 723 | ||
NM_001033716.1 | NP_001028888.1 | 8 | 654 | |||
NM_053295.2 | NP_445747.2 | 10 | 837 | |||
XM_006231698.2 | XP_006231760.1 | 10 | 1245 | |||
XM_006231699.2 | XP_006231761.1 | 9 | 970 | |||
GenBank mRNA | AF346597.1 | - | 6 | 335 | ||
AF346598.1 | - | 8 | 518 | |||
BC091239.1 | - | 9 | 723 | 7324026 | ||
DQ186624.1 | - | 10 | 1031 | |||
DQ186625.1 | - | 8 | 848 | |||
DQ186626.1 | - | 9 | 962 | |||
DQ186627.1 | - | 9 | 917 | |||
DQ186628.1 | - | 8 | 878 | |||
DQ186629.1 | - | 7 | 809 | |||
DQ287975.1 | - | 3 | 709 | |||
Y13587.1 | - | 7 | 416 | |||
Y13588.1 | - | 8 | 530 | |||
Y13589.1 | - | 6 | 347 | |||
Y13591.1 | - | 7 | 416 |
