siRNA Details
| Gene Symbol: Fgf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012952.1 | NP_037084.1 | 1 | 175 | ||
| GenBank mRNA | BC078699.1 | - | 778 | |||
| D14839.1 | - | 1 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf9| Gene Symbol: Fgf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012952.1 | NP_037084.1 | 1 | 175 | ||
| GenBank mRNA | BC078699.1 | - | 778 | |||
| D14839.1 | - | 1 | 194 | |||