siRNA Details
| Gene Symbol: Hrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012965.3 | NP_037097.3 | 1 | 727 | ||
| XM_008771486.1 | XP_008769708.1 | 1 | 2034 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrh2| Gene Symbol: Hrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012965.3 | NP_037097.3 | 1 | 727 | ||
| XM_008771486.1 | XP_008769708.1 | 1 | 2034 | |||
Set Membership: |
|