siRNA Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105719.1 | NP_001099189.1 | 4 | 813 | ||
| GenBank mRNA | AB684453.1 | - | 4 | 773 | ||
| FQ233861.1 | - | 847 | ||||
| X53052.1 | - | 4 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mip| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105719.1 | NP_001099189.1 | 4 | 813 | ||
| GenBank mRNA | AB684453.1 | - | 4 | 773 | ||
| FQ233861.1 | - | 847 | ||||
| X53052.1 | - | 4 | 762 | |||