siRNA Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013028.1 | NP_037160.1 | 5 | 688 | ||
| GenBank mRNA | AJ002258.1 | - | 5 | 673 | ||
| AJ002259.1 | - | 5 | 688 | |||
| AJ002260.1 | - | 4 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shox2| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013028.1 | NP_037160.1 | 5 | 688 | ||
| GenBank mRNA | AJ002258.1 | - | 5 | 673 | ||
| AJ002259.1 | - | 5 | 688 | |||
| AJ002260.1 | - | 4 | 625 | |||