siRNA Details
| Gene Symbol: Ywhah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013052.1 | NP_037184.1 | 2 | 639 | ||
| GenBank mRNA | BC081825.1 | - | 2 | 664 | 7192904 | |
| D17445.1 | - | 2 | 639 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ywhah| Gene Symbol: Ywhah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013052.1 | NP_037184.1 | 2 | 639 | ||
| GenBank mRNA | BC081825.1 | - | 2 | 664 | 7192904 | |
| D17445.1 | - | 2 | 639 | |||