siRNA Details
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013053.1 | NP_037185.1 | 2, 3 | 379 | ||
GenBank mRNA | BC062409.1 | - | 1, 2 | 349 | 5623850 | |
D17614.1 | - | 2, 3 | 379 | |||
FQ218679.1 | - | 1, 2 | 139 |
