siRNA Details
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013065.2 | NP_037197.1 | 5 | 831 | ||
XM_008764505.1 | XP_008762727.1 | 6 | 769 | |||
GenBank mRNA | BC062033.1 | - | 5 | 831 | 5623630 | |
D90164.1 | - | 5 | 611 | |||
FQ212294.1 | - | 6 | 769 | |||
S78218.1 | - | 5 | 598 |
