siRNA Details
| Gene Symbol: Hoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013075.1 | NP_037207.1 | 1 | 295 | ||
| XM_008773708.1 | XP_008771930.1 | 1 | 411 | |||
| GenBank mRNA | U93092.1 | - | 1 | 295 | ||
| U93093.1 | - | 1 | 280 | |||
| Gene Symbol: NEWGENE_2812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762937.1 | XP_008761159.1 | 1 | 411 | ||
| XM_008762938.1 | XP_008761160.1 | 1 | 411 | |||

Hoxa1
Genomic Map