siRNA Details
| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177593.1 | NP_001171064.1 | 14 | 1826 | ||
| NM_013088.2 | NP_037220.2 | 14 | 1814 | |||
| GenBank mRNA | U09307.1 | - | 14 | 1739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn11| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177593.1 | NP_001171064.1 | 14 | 1826 | ||
| NM_013088.2 | NP_037220.2 | 14 | 1814 | |||
| GenBank mRNA | U09307.1 | - | 14 | 1739 | ||