siRNA Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 4 | 653 | ||
| GenBank mRNA | AF110461.2 | - | 4 | 345 | ||
| L27057.1 | - | 4 | 653 | |||
| L36467.1 | - | 543 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde4a| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 4 | 653 | ||
| GenBank mRNA | AF110461.2 | - | 4 | 345 | ||
| L27057.1 | - | 4 | 653 | |||
| L36467.1 | - | 543 | ||||