siRNA Details
| Gene Symbol: Pde4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013101.3 | NP_037233.3 | 9 | 1220 | ||
| GenBank mRNA | AF110461.2 | - | 9 | 912 | ||
| L27057.1 | - | 9 | 1220 | |||
| L27062.1 | - | 3 | 750 | |||
| L36467.1 | - | 1110 | ||||
| M25348.1 | - | 2 | 180 | |||
| M26715.1 | - | 3 | 484 | |||
| M26716.1 | - | 3 | 388 | |||
| M28411.1 | - | 2 | 180 | |||

Pde4a
Genomic Map