siRNA Details
| Gene Symbol: Igfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013144.1 | NP_037276.1 | 1 | 519 | ||
| GenBank mRNA | BC078889.1 | - | 1 | 501 | 7129185 | |
| M58634.1 | - | 1 | 519 | |||
| M89791.1 | - | 1 | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp1| Gene Symbol: Igfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013144.1 | NP_037276.1 | 1 | 519 | ||
| GenBank mRNA | BC078889.1 | - | 1 | 501 | 7129185 | |
| M58634.1 | - | 1 | 519 | |||
| M89791.1 | - | 1 | 480 | |||