siRNA Details
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013185.3 | NP_037317.2 | 11 | 1423 | ||
GenBank mRNA | BC078890.1 | - | 11 | 1423 | 7121407 | |
M83666.1 | - | 11 | 1345 | |||
S74141.1 | - | 11 | 1345 | |||
X62345.1 | - | 11 | 1374 |
