siRNA Details
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013185.3 | NP_037317.2 | 8 | 968 | ||
GenBank mRNA | BC078890.1 | - | 8 | 968 | 7121407 | |
X62345.1 | - | 8 | 919 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013185.3 | NP_037317.2 | 8 | 968 | ||
GenBank mRNA | BC078890.1 | - | 8 | 968 | 7121407 | |
X62345.1 | - | 8 | 919 |