siRNA Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013197.1 | NP_037329.1 | 7 | 1143 | ||
| GenBank mRNA | BC081857.1 | - | 1164 | 7133382 | ||
| D86297.1 | - | 7 | 1143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alas2| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013197.1 | NP_037329.1 | 7 | 1143 | ||
| GenBank mRNA | BC081857.1 | - | 1164 | 7133382 | ||
| D86297.1 | - | 7 | 1143 | |||