siRNA Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147216.1 | NP_671749.1 | 3 | 624 | ||
| GenBank mRNA | AJ496570.1 | - | 3 | 624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a2| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147216.1 | NP_671749.1 | 3 | 624 | ||
| GenBank mRNA | AJ496570.1 | - | 3 | 624 | ||