siRNA Details
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153315.2 | NP_695227.2 | 6 | 745 | ||
GenBank mRNA | AB062758.1 | - | 6 | 561 | ||
BC070961.1 | - | 6 | 745 | 7099044 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153315.2 | NP_695227.2 | 6 | 745 | ||
GenBank mRNA | AB062758.1 | - | 6 | 561 | ||
BC070961.1 | - | 6 | 745 | 7099044 |