siRNA Details
| Gene Symbol: Gzmf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153466.1 | NP_703196.1 | 3 | 315 | ||
| GenBank mRNA | U57063.1 | - | 3 | 315 | ||
| Gene Symbol: LOC100910060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770727.1 | XP_008768949.1 | 2 | 227 | ||

Gzmf
Genomic Map