siRNA Details
| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172223.2 | NP_757377.1 | 2, 3 | 215 | ||
| GenBank mRNA | AY151834.1 | - | 2, 3 | 193 | ||
| BC081699.1 | - | 2, 3 | 215 | 7193564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pxmp4| Gene Symbol: Pxmp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172223.2 | NP_757377.1 | 2, 3 | 215 | ||
| GenBank mRNA | AY151834.1 | - | 2, 3 | 193 | ||
| BC081699.1 | - | 2, 3 | 215 | 7193564 | ||