siRNA Details
| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172335.2 | NP_758838.2 | 2 | 238 | ||
| GenBank mRNA | AB051391.1 | - | 2 | 197 | ||
| BC072474.1 | - | 2 | 238 | 7100875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2a| Gene Symbol: Gm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172335.2 | NP_758838.2 | 2 | 238 | ||
| GenBank mRNA | AB051391.1 | - | 2 | 197 | ||
| BC072474.1 | - | 2 | 238 | 7100875 | ||