siRNA Details
| Gene Symbol: Rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004075.1 | NP_001004075.1 | 2 | 259 | ||
| XM_006249360.2 | XP_006249422.1 | 2 | 792 | |||
| XM_006249361.2 | XP_006249423.1 | 3 | 469 | |||
| XM_008769214.1 | XP_008767436.1 | 2 | 177 | |||
| GenBank mRNA | AY157968.1 | - | 1 | 164 | ||
| BC079044.1 | - | 2 | 259 | 7115027 | ||

Rnf34
Genomic Map