siRNA Details
| Gene Symbol: Rnf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004075.1 | NP_001004075.1 | 2 | 115 | ||
| XM_006249360.2 | XP_006249422.1 | 2 | 648 | |||
| XM_006249361.2 | XP_006249423.1 | 3 | 325 | |||
| XM_008769214.1 | XP_008767436.1 | 2 | 33 | |||
| GenBank mRNA | AY157968.1 | - | 1 | 20 | ||
| BC079044.1 | - | 2 | 115 | 7115027 | ||

Rnf34
Genomic Map