siRNA Details
| Gene Symbol: Wfdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173109.1 | NP_775132.1 | 3 | 307 | ||
| XM_008762473.1 | XP_008760695.1 | 3 | 312 | |||
| GenBank mRNA | AJ515239.1 | - | 3 | 307 | ||
| AJ515241.1 | - | 2 | 127 | |||
| FQ222047.1 | - | 1 | 65 | |||

Wfdc2
Genomic Map