siRNA Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134350.2 | NP_599177.2 | 5, 6 | 712 | ||
| XM_006248150.2 | XP_006248212.1 | 7, 8 | 856 | |||
| XM_008768568.1 | XP_008766790.1 | 7, 8 | 790 | |||
| GenBank mRNA | BC099808.1 | - | 5, 6 | 712 | 7938638 | |
| X52712.1 | - | 5, 6 | 787 | |||
| X52713.1 | - | 4, 5 | 592 | |||

Mx2
Genomic Map