siRNA Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134350.2 | NP_599177.2 | 10 | 1301 | ||
| XM_006248150.2 | XP_006248212.1 | 12 | 1445 | |||
| XM_008768568.1 | XP_008766790.1 | 12 | 1379 | |||
| GenBank mRNA | BC099808.1 | - | 10 | 1301 | 7938638 | |
| X52712.1 | - | 10 | 1376 | |||
| X52713.1 | - | 9 | 1181 | |||

Mx2
Genomic Map