siRNA Details
Gene Symbol: Fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173308.2 | NP_775430.2 | 2 | 1106 | ||
GenBank mRNA | AJ535753.1 | - | 2 | 1131 | ||
BC079316.1 | - | 2 | 1106 | 7135022 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fut11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173308.2 | NP_775430.2 | 2 | 1106 | ||
GenBank mRNA | AJ535753.1 | - | 2 | 1131 | ||
BC079316.1 | - | 2 | 1106 | 7135022 |