siRNA Details
| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173311.2 | NP_775433.2 | 7 | 606 | ||
| GenBank mRNA | AB092812.1 | - | 7 | 508 | ||
| BC097931.1 | - | 7 | 606 | 7389090 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skap1| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173311.2 | NP_775433.2 | 7 | 606 | ||
| GenBank mRNA | AB092812.1 | - | 7 | 508 | ||
| BC097931.1 | - | 7 | 606 | 7389090 | ||