siRNA Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191588.1 | NP_001178517.1 | 2 | 217 | ||
| XM_006245784.1 | XP_006245846.1 | 2 | 514 | |||
| Gene Symbol: LOC100910915 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245828.2 | XP_006245890.1 | 2 | 508 | ||

Nmral1
Genomic Map