siRNA Details
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102364.1 | NP_001095834.1 | 2 | 143 | ||
XM_008767511.1 | XP_008765733.1 | 3 | 374 | |||
GenBank mRNA | BC168250.1 | - | 2 | 143 | 5598970 |
