siRNA Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011890.1 | NP_001011890.1 | 3 | 343 | ||
XM_006245931.2 | XP_006245993.1 | 3 | 787 | |||
XM_006245932.2 | XP_006245994.1 | 4 | 486 | |||
XM_006245933.2 | XP_006245995.1 | 2 | 282 | |||
GenBank mRNA | BC079014.1 | - | 3 | 343 | 7113485 |
