siRNA Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011890.1 | NP_001011890.1 | 4, 5 | 593 | ||
XM_006245931.2 | XP_006245993.1 | 4, 5 | 1037 | |||
XM_006245932.2 | XP_006245994.1 | 5, 6 | 736 | |||
XM_006245933.2 | XP_006245995.1 | 3, 4 | 532 | |||
GenBank mRNA | BC079014.1 | - | 4, 5 | 593 | 7113485 |
