siRNA Details
| Gene Symbol: Hbz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172845.1 | NP_001166316.1 | 3 | 465 | ||
| GenBank mRNA | FQ221080.1 | - | 3 | 473 | ||
| FQ221398.1 | - | 3 | 473 | |||
| FQ222010.1 | - | 3 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hbz| Gene Symbol: Hbz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172845.1 | NP_001166316.1 | 3 | 465 | ||
| GenBank mRNA | FQ221080.1 | - | 3 | 473 | ||
| FQ221398.1 | - | 3 | 473 | |||
| FQ222010.1 | - | 3 | 476 | |||