siRNA Details
Gene Symbol: Olr1386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214834.1 | NP_999999.1 | 1 | 13 | ||
XM_006246176.2 | XP_006246238.1 | 2 | 68 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214834.1 | NP_999999.1 | 1 | 13 | ||
XM_006246176.2 | XP_006246238.1 | 2 | 68 |