siRNA Details
| Gene Symbol: Dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008507.1 | NP_001008507.1 | 6 | 659 | ||
| XM_006246472.2 | XP_006246534.1 | 7 | 867 | |||
| XM_006246473.2 | XP_006246535.1 | 6 | 1528 | |||
| XM_006246474.2 | XP_006246536.1 | 6 | 996 | |||
| XM_006246475.2 | XP_006246537.1 | 5 | 679 | |||
| XM_008767786.1 | XP_008766008.1 | 6 | 855 | |||
| GenBank mRNA | BC086453.1 | - | 6 | 659 | 7305216 | |
| EF445633.1 | - | 7 | 771 | |||

Dhrs7b
Genomic Map