siRNA Details
| Gene Symbol: Elp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001718.1 | NP_001001718.1 | 5, 6 | 859 | ||
| XM_006246639.2 | XP_006246701.1 | 6, 7 | 1059 | |||
| XM_006246640.1 | XP_006246702.1 | 4, 5 | 368 | |||
| GenBank mRNA | AY623900.1 | - | 5, 6 | 859 | ||
| BC158569.1 | - | 6, 7 | 1062 | 9101477 | ||

Elp5
Genomic Map