siRNA Details
| Gene Symbol: Txndc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105805.1 | NP_001099275.1 | 3 | 320 | ||
| GenBank mRNA | BC159408.1 | - | 3 | 248 | 8362552 | |
| FQ211488.1 | - | 3 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txndc17| Gene Symbol: Txndc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105805.1 | NP_001099275.1 | 3 | 320 | ||
| GenBank mRNA | BC159408.1 | - | 3 | 248 | 8362552 | |
| FQ211488.1 | - | 3 | 291 | |||