siRNA Details
| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105819.1 | NP_001099289.1 | 3 | 508 | ||
| XM_006246980.2 | XP_006247042.1 | 2 | 545 | |||
| XM_006246981.1 | XP_006247043.1 | 3 | 1234 | |||
| XM_006246982.2 | XP_006247044.1 | 3 | 570 | |||
| XM_008767963.1 | XP_008766185.1 | 3 | 570 | |||

Rhbdl3
Genomic Map