siRNA Details
| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105819.1 | NP_001099289.1 | 1, 2 | 370 | ||
| XM_006246982.2 | XP_006247044.1 | 1, 2 | 432 | |||
| XM_008767963.1 | XP_008766185.1 | 1, 2 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhbdl3| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105819.1 | NP_001099289.1 | 1, 2 | 370 | ||
| XM_006246982.2 | XP_006247044.1 | 1, 2 | 432 | |||
| XM_008767963.1 | XP_008766185.1 | 1, 2 | 432 | |||