siRNA Details
| Gene Symbol: Krt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004022.2 | NP_001004022.1 | 2 | 557 | ||
| GenBank mRNA | BC101868.1 | - | 2 | 557 | 7938504 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt15| Gene Symbol: Krt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004022.2 | NP_001004022.1 | 2 | 557 | ||
| GenBank mRNA | BC101868.1 | - | 2 | 557 | 7938504 | |