siRNA Details
| Gene Symbol: Krt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212545.2 | NP_997710.1 | 8 | 1293 | ||
| GenBank mRNA | BC100058.1 | - | 8 | 1293 | 7383488 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt17| Gene Symbol: Krt17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212545.2 | NP_997710.1 | 8 | 1293 | ||
| GenBank mRNA | BC100058.1 | - | 8 | 1293 | 7383488 | |