siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199082.2 | NP_954513.2 | 4 | 601 | ||
| XM_006247879.2 | XP_006247941.1 | 5 | 757 | |||
| XM_006247880.2 | XP_006247942.1 | 6 | 1581 | |||
| XM_006247881.2 | XP_006247943.1 | 5 | 1298 | |||
| XM_006247882.2 | XP_006247944.1 | 4 | 694 | |||
| XM_006247884.2 | XP_006247946.1 | 5 | 1519 | |||
| GenBank mRNA | BC061794.1 | - | 4 | 596 | 5597864 | |
| DY309177.1 | - | 3 | 689 | |||

Sectm1b
Genomic Map